missing translation for 'onlineSavingsMsg'
Learn More

PCPE-1 Rat anti-Human, Clone: 261730, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB2627

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459585

  • £279.85 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

PCPE-1 Monoclonal antibody MAB2627 Specifically detects PCPE-1 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

PCPE-1/PCOLCE
261730
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PCOLCE
Mouse myeloma cell line NS0-derived recombinant human PCPE-1, Gln26-Asp449 (predicted), Accession # Q15113
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5118
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Q15113
PCOLCE, PCPE, PCPE1, PCPE-1, PCPE1Type I procollagen COOH-terminal proteinase enhancer, procollagen C-endopeptidase enhancer, procollagen C-endopeptidase enhancer 1, Procollagen COOH-terminal proteinase enhancer 1, Procollagen C-proteinase enhancer 1, procollagen, type 1, COOH-terminal proteinase enhancer, Type 1 procollagen C-proteinase enhancer protein
Rat
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human PCPE-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse PCPE-1 is observed.
Human
Videos

For Research Use Only