missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDHA12 Monoclonal Antibody (1D3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA522259
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: LTGSVQIQIT VLDVNDNGPA FDKPSYKVVL SENVQNDTRV IQLNASDPDE GLNGEISYGI KMILPVSEKC MFSINPDTGE IRIYGELDFE ENNAYEIQVN AIDKGI
PCDHA12 Monoclonal antibody specifically detects PCDHA12 in Human samples. It is validated for ELISA.Specifications
PCDHA12 | |
Monoclonal | |
0.25 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9UN75 | |
PCDHA12 | |
PCDHA12 (NP_061726, 222 a.a. ∽ 327 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA | |
1D3 | |
Unconjugated | |
PCDHA12 | |
cadherin-related neuronal receptor 5; Cnr5; Crnr5; KIAA0345-like 2; MGC138485; MGC141932; PCDHA12; Pcdha13; PCDH-ALPHA12; PCDH-alpha-12; protocadherin alpha 12; protocadherin alpha 13; protocadherin alpha-12; rCNRv12 | |
Mouse | |
Affinity chromatography | |
RUO | |
56137 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |