Learn More
PBX2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541437
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: PGGYGDNLGG GQMYSPREMR ANGSWQEAVT PSSVTSPTEG PGSVHSDTSN Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
PBX2 Polyclonal antibody specifically detects PBX2 in Human samples. It is validated for Western Blot.Specifications
PBX2 | |
Polyclonal | |
Unconjugated | |
PBX2 | |
AU043397; G17; homeobox 12; homeobox protein PBX2; HOX12; PBX homeobox 2; Pbx2; PBX2MHC; PBX2P1; pbxy; pbxy homeodomain protein; pre B cell leukemia homeobox 2; pre B-cell leukemia transcription factor 2; pre-B-cell leukemia homeobox 2; pre-B-cell leukemia homeobox 2 pseudogene 1; pre-B-cell leukemia homeobox 3; pre-B-cell leukemia transcription factor 2; pre-B-cell leukemia transcription factor 2; pre-B-cell leukemia homeobox 2; pre-B-cell leukemia transcription factor y; Protein G17; wu:fb73g03; XXbac-BPG300A18.13 | |
Rabbit | |
Protein A | |
RUO | |
5089 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P40425 | |
PBX2 | |
Synthetic peptide directed towards the C-terminal of human PBX2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |