missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine Parvovirus Monoclonal Antibody (H69D), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17014
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-7014 detects the canine Parvovirus from infected samples. MA1-7014 has been successfully used in ELISA procedures. The MA1-7014 immunogen is canine Parvovirus lysate.
Canine Parvovirus Monoclonal antibody specifically detects Canine Parvovirus in Virus samples. It is validated for ELISA.Specifications
Canine Parvovirus | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
ELISA | |
H69D | |
Unconjugated | |
CPV; parvo | |
Canine Parvovirus lysate. | |
1 mL | |
Primary | |
4° C | |
Liquid |