Learn More
Pappalysin-2/PAPP-A2 Mouse anti-Human, Clone: 242011, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1668
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448815
  • £416.01 / 500µg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
Pappalysin-2/PAPP-A2 Monoclonal antibody MAB1668 Specifically detects Pappalysin-2/PAPP-A2 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Pappalysin-2/PAPP-A2
242011
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PAPPA2
Mouse myeloma cell line NS0-derived recombinant human Pappalysin-2/PAPP-A2 Ser234-Cys1396 Accession # Q9BXP8
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
60676
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Q9BXP8
EC 3.4.24.-, PAPPA2, PAPP-A2, PAPP-A2pappalysin-2, pappalysin 2, Pappalysin2, PAPPE, PAPP-E, PLAC3, placenta-specific 3, pregnancy-associated plasma preproprotein-A2, Pregnancy-associated plasma protein A2, Pregnancy-associated plasma protein E1
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Pappalysin-2/PAPP-A2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human PAPP-A is observed.