Learn More
PAK4, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PAK4.
Brand: Abnova H00010298-A01.50uL
Additional Details : Weight : 0.00010kg
Description
PAK proteins, a family of serine/threonine p21-activating kinases, include PAK1, PAK2, PAK3 and PAK4. PAK proteins are critical effectors that link Rho GTPases to cytoskeleton reorganization and nuclear signaling. They serve as targets for the small GTP binding proteins Cdc42 and Rac and have been implicated in a wide range of biological activities. PAK4 interacts specifically with the GTP-bound form of Cdc42Hs and weakly activates the JNK family of MAP kinases. PAK4 is a mediator of filopodia formation and may play a role in the reorganization of the actin cytoskeleton. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq
Sequence: KTIVRGSKGAKDGALTLLLDEFENMSVTRSNSLRRDSPPPPARARQENGMPEKPPGPRSPQREPQRVSHEQFRAALQLVVDPGDPRSYLDSpecifications
PAK4 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant PAK4. | |
PAK4 | |
PAK4 | |
PAK4 (AAH02921, 68 a.a. ∼ 157 a.a) partial recombinant protein with GST tag. | |
RUO | |
10298 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
BC002921 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Liquid |