missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PACSIN3 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA517995
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
PACSIN3 Polyclonal antibody specifically detects PACSIN3 in Human samples. It is validated for Western Blot.Specifications
PACSIN3 | |
Polyclonal | |
Unconjugated | |
PACSIN3 | |
4921507A02Rik; 6330413E15Rik; AW413130; EGK_06300; Endophilin I; PACSIN3; Protein kinase C and casein kinase II substrate protein 3; protein kinase C and casein kinase substrate in neurons 3; protein kinase C and casein kinase substrate in neurons protein 3; SDPIII; SH3 domain-containing protein 6511; syndapin III | |
Goat | |
Antigen affinity chromatography | |
RUO | |
29763 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9UKS6 | |
PACSIN3 | |
Synthetic peptide sequence (APEEDAGGEALGGS) corresponding to the N-terminus amino acids of PACSIN3 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |