missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fyn Monoclonal Antibody (Fyn-1S), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 137800
Code : X3
Additional Details : Weight : 0.50000kg
Description
FYN is a SRC-related kinase expressed in brain, skeletal muscle and adipose tissues. Fyn functions in a large number of pathways and has been shown to play a role in memory. FYN is a member of the protein-tyrosine kinase oncogene family. It encodes a membrane-associated tyrosine kinase that has been implicated in the control of cell growth. The protein associates with the p85 subunit of phosphatidylinositol 3-kinase and interacts with the fyn-binding protein. Alternatively spliced transcript variants encoding distinct isoforms exist.Specifications
Fyn | |
Monoclonal | |
0.5 mg/mL | |
PBS with 1% BSA and 0.1% sodium azide | |
P06241, P39688 | |
Fyn | |
A fusion protein consisting of amino acids 1-206 of murine p59fyn. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
Fyn-1S | |
Unconjugated | |
Fyn | |
AI448320; AW552119; C syn protooncogene; c-syn protooncogene; EC 2.7.10.2; Fyn; fyn {ECO:0000312; FYN oncogene related to SRC, FGR, YES; Fyn proto-oncogene; FYN proto-oncogene, Src family tyrosine kinase; kinase Fyn; MGC45350; OKT3-induced calcium influx regulator; OTTHUMP00000017916; OTTHUMP00000017917; P59 p59-Fyn; p59-Fyn; Proto-oncogene c-Fyn; Protooncogene Syn; proto-oncogene Syn; proto-oncogene tyrosine-protein kinase Fyn; RGD:2641}; RP1-66H14.1; SLK; Src Kinase p59; Src like kinase; src/yes-related novel; src-like kinase; SYN; tyrosine kinase p59fyn(T); tyrosine-protein kinase Fyn | |
Mouse | |
Affinity chromatography | |
RUO | |
14360, 2534 | |
-20°C | |
Liquid |