Learn More
IFIT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA3845
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA3-845 detects ISG54 (p54) from human and mouse samples. PA3-845 has been successfully used in ICC/IF and in Western blot applications to detect a 39-58 kDa band. This antibody may produce multiple background bands, and it is recommended to run samples slowly on an 8% gel to achieve proper separation.
IFIT2 Polyclonal antibody specifically detects IFIT2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
IFIT2 | |
Polyclonal | |
Unconjugated | |
Ifit2 | |
AV302338; cig42; CIG-42; G10P2; Garg39; GARG-39; Glucocorticoid-attenuated response gene 39 protein; IFI54; IFI-54; IFI-54K; Ifit2; IFIT-2; interferon induced protein with tetratricopeptide repeats 2; Interferon, alpha-inducible protein (MW 54kD); interferon-induced 54 kDa protein; interferon-induced protein 54; interferon-induced protein with tetratricopeptide repeats 2; ISG54; ISG-54 K; ISG-54K; P54 | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
Conc. Not Determined | |
whole serum with 0.05% sodium azide | |
P09913, Q64112 | |
Ifit2 | |
Bacterially expressed full-length mouse p54 protein | |
RUO | |
15958, 294091, 3433 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |