missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p53 Monoclonal Antibody (PAb 240), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515244
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-15244 targets p53 in FACS, IHC (P), IP, and WB applications and shows reactivity with Human samples. The MA5-15244 immunogen is mouse p53 protein corresponding to aa14-289.
p53 Monoclonal antibody specifically detects p53 in Human, Mouse, Rhesus Monkey samples. It is validated for ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
p53 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P02340, P04637, P56424 | |
Tp53, Trp53 | |
Murine p53 protein corresponding to aa14-289 | |
500 μL | |
Primary | |
Human, Mouse, Rhesus Monkey | |
Antibody | |
IgG1 |
ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
PAb 240 | |
Unconjugated | |
Tp53 | |
Antigen NY-CO-13; bbl; BCC7; bfy; bhy; cellular tumor antigen p53; Cys 51 Stop; EGK_08142; FLJ92943; HGNC11998; I79_002739; LFS1; Li-Fraumeni syndrome; mutant p53; mutant tumor protein 53; OTTMUSP00000006194; p44; p53; p53 cellular tumor antigen; p53 protein; p53 tumor suppressor; p53 tumor suppressor phosphoprotein; phosphoprotein p53; Tp53; tp53.L; transformation related protein 53; transformation-related protein 53; Trp248; Trp53; tumor protein 53; tumor protein p53; tumor protein p53 (Li-Fraumeni syndrome); tumor protein p53 L homeolog; tumor suppressor p53; tumor suppressor p53 phosphoprotein; tumor suppressor protein p53; tumor supressor p53; Tumour Protein p53; XELAEV_180196761mg; Xp53; Xrel3 | |
Mouse | |
Protein G | |
RUO | |
22059, 7157, 716170 | |
4° C | |
Liquid |