Learn More
p53 Mouse anti-Human, Clone: 184727, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB13551
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448575
  • £277.83 / 100µg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
p53 Monoclonal antibody MAB13551 Specifically detects p53 in Human samples. It is validated for Flow Cytometry (Intracellular Staining)
Specifications
Specifications
p53
184727
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TP53
E. coli-derived recombinant human p53 Asp7-Asp393 Accession # P04637
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7157
Reconstitute at 0.5 mg/mL in sterile PBS.
Flow Cytometry, CyTOF
LYOPH
Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
P04637
Antigen NY-CO-13, BCC7, FLJ92943, LFS1, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, TP53, transformation-related protein 53, TRP53, tumor protein p53, Tumor suppressor p53
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human p53.