missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p40 Monoclonal Antibody (4C9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA520977
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: YHTEEQHWTL LKFDTLLPPG RLDHSMCIIP WPVTCASEKE DSNSLTLNHE AEKEDSADKV MSHSGDSHEE SQTATLLCLV FGGMNTEGEI YDDCIVTVVD
p40 Monoclonal antibody specifically detects p40 in Human samples. It is validated for ELISA, Western Blot.Specifications
p40 | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q7Z6M1 | |
Rabepk | |
RABEPK (NP_005824.2, 273 a.a. ∽ 372 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
4C9 | |
Unconjugated | |
Rabepk | |
40 kDa Rab9 effector protein; 8430412M01Rik; 9530020d24rik; AV073337; bA65N13.1; C87311; p40; Rab9 effector p40; rab9 effector protein with kelch motifs; RAB9P40; RABEPK; RGD1310612 | |
Mouse | |
Affinity chromatography | |
RUO | |
10244 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |