missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p38 MAPK alpha Monoclonal Antibody (G.876.7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515204
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with either JNK/SAPK or p42/44 MAPK or other isoforms of p38.
p38 MAPK alpha Monoclonal antibody specifically detects p38 MAPK alpha in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
p38 MAPK alpha | |
Monoclonal | |
Unconjugated | |
MAPK14 | |
186F5S; anon-sts23; BG:DS00797.3; CG7393; CG7393-PA; CRK1; CSAID-binding protein; Csaids binding protein; CSBP; Csbp1; Csbp2; CSPB1; cytokine suppressive anti-inflammatory drug binding protein; cytokine suppressive anti-inflammatory drug binding protein 1; cytokine suppressive anti-inflammatory drug-binding protein; cytokine-supressive anti-inflammatory drug binding protein; Dm p38b; Dmel\CG7393; Dmel_CG7393; Dmp38b; Dp38; D-p38; D-p38 MAPK; Dp38b; D-p38b; EC 2.7.11.24; ERK1; ERT2; ESTS:186F5S; etID18993.8; Exip; fk28c03; hm:zeh1243; Hog; I79_016605; LD24658p; MAP 14; MAP kinase 1; MAP kinase 11; MAP kinase 14; MAP kinase 14A; MAP kinase 2; MAP kinase Mxi2; MAP kinase p38; MAP kinase p38 alpha; MAP kinase p38 beta; MAP kinase p38a; MAP kinase p38alpha; MAP kinase p38b; MAPK; MAPK 11; MAPK 14; MAPK 14A; MAPK1 antibody; Mapk11; Mapk14; mapk14a; MAX-interacting protein 2; mitogen activated protein kinase 11; mitogen activated protein kinase 14; mitogen-activated protein kinase 11; Mitogen | |
Mouse | |
Affinity chromatography | |
RUO | |
1432, 26416, 81649 | |
-20°C | |
Liquid |
Western Blot | |
G.876.7 | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P47811, P70618, Q16539 | |
MAPK14 | |
Recombinant p38 MAPK fusion protein | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |