Learn More
P2Y2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530113
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, Jurkat, Raji, mouse brain, rat brain. Predicted reactivity: Human (99%), Mouse (84%), Rat (85%), Pig (82%), Bovine (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
P2Y2 Polyclonal antibody specifically detects P2Y2 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
P2Y2 | |
Polyclonal | |
Unconjugated | |
P2RY2 | |
ATP receptor; HP2U; P2RU1; P2ry2; P2U; P2U nucleotide receptor; P2U purinoceptor 1; P2U receptor 1; P2U1; P2UR; P2Y ATP receptor 2; P2Y purinoceptor 2; P2Y2; P2Y2R; P2Y-like receptor; Purinergic receptor; purinergic receptor P2Y, G-protein coupled 2; purinergic receptor P2Y, G-protein coupled, 2; purinergic receptor P2Y2; purinoceptor P2Y2; R12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18442, 5029 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
1.12 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P35383, P41231 | |
P2RY2 | |
Recombinant protein fragment corresponding to a region within amino acids 164 and 327 of Human P2Y2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |