Learn More
P2Y2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA14517
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-4517 detects P2Y purinoceptor 2 in human, rat and mouse samples. PA1-4517 has been successfully used in Western blot and Immunohistochemistry procedures. PA1-4517 immunogen is a synthetic peptide comprising of internal residues of the human P2Y2 protein. Reacts with rat and mouse P2Y2. Maintain at 4oC for 3 months or at -20oC for longer periods. Avoid freeze/thaw cycles.
P2Y2 Polyclonal antibody specifically detects P2Y2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
P2Y2 | |
Polyclonal | |
Unconjugated | |
P2RY2 | |
ATP receptor; HP2U; P2RU1; P2ry2; P2U; P2U nucleotide receptor; P2U purinoceptor 1; P2U receptor 1; P2U1; P2UR; P2Y ATP receptor 2; P2Y purinoceptor 2; P2Y2; P2Y2R; P2Y-like receptor; Purinergic receptor; purinergic receptor P2Y, G-protein coupled 2; purinergic receptor P2Y, G-protein coupled, 2; purinergic receptor P2Y2; purinoceptor P2Y2; R12 | |
Rabbit | |
Affinity chromatography | |
RUO | |
18442, 29597, 5029 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P35383, P41231, P41232 | |
P2RY2 | |
Synthetic peptide corresponding to residues of the human P2Y2 protein. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |