missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534084
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Gibbon, Marmoset (93%) Panda, Dog (87%).
P2Y13 Polyclonal antibody specifically detects P2Y13 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
P2Y13 | |
Polyclonal | |
Unconjugated | |
P2RY13 | |
2010001L06Rik; FKSG77; G protein-coupled receptor 68; G protein-coupled receptor 86; GPCR1; GPR86; GPR94; G-protein coupled receptor 86; G-protein coupled receptor 94; P2ry13; P2Y purinoceptor 13; P2Y13; P2yr13; purinergic receptor P2Y G-protein coupled 13; purinergic receptor P2Y, G-protein coupled 13; purinergic receptor P2Y, G-protein coupled, 13; purinergic receptor P2Y13; SP174; Vpurinergic receptor P2Y, G-protein coupled 13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
53829 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9BPV8 | |
P2RY13 | |
Synthetic 15 amino acid peptide from N-terminal extracellular domain of human P2RY13 / P2Y13. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |