missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534080
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Panda, Dog, Horse, Rabbit (100%) Bat, Pig, Platypus (94%).
P2Y12 Polyclonal antibody specifically detects P2Y12 in Canine, Human, Non-human Primate, Rabbit samples. It is validated for Immunohistochemistry (Paraffin).Specifications
P2Y12 | |
Polyclonal | |
Unconjugated | |
P2ry12 | |
2900079B22Rik; 4921504D23Rik; ADP-glucose receptor; ADPG-R; BDPLT8; Gi-coupled ADP receptor HORK3; G-protein coupled receptor SP1999; HORK3; P2RY12; P2T(AC); P2Y purinoceptor 12; P2Y(12)R; P2Y(AC); P2Y(ADP); P2Y(cyc); P2y12; P2Y12 platelet ADP receptor; P2YR12; purinergic receptor P2RY12; purinergic receptor P2Y, G-protein coupled 12; purinergic receptor P2Y, G-protein coupled, 12; purinergic receptor P2Y12; putative G-protein coupled receptor; SP1999 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100345006, 442958, 64805 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9H244 | |
P2ry12 | |
Synthetic 16 amino acid peptide from 2nd extracellular domain of human P2RY12 / P2Y12. | |
50 μg | |
Primary | |
Canine, Human, Non-human Primate, Rabbit | |
Antibody | |
IgG |