missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X4 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA537880
Code : NEW
Additional Details : Weight : 0.00900kg
Description
This antibody is predicted to react with mouse, rat, dog, pig and cow based on sequence homology.
P2X4 Polyclonal antibody specifically detects P2X4 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
P2X4 | |
Polyclonal | |
Unconjugated | |
P2RX4 | |
AI504491; ATP receptor; ATP-gated cation channel protein; AW555605; D5Ertd444e; ionotropic purinergic receptor; P2 purinergic receptor subtype; P2RX4; P2X purinoceptor 4; P2X receptor, subunit 4; P2X4; P2X4 purinoceptor; P2x4 receptor; P2X4R; Purinergic receptor; purinergic receptor P2X 4; purinergic receptor P2X, ligand gated ion channel, 4; purinergic receptor P2X, ligand-gated ion channel 4; purinergic receptor P2X, ligand-gated ion channel, 4; purinergic receptor P2X4; purinergic receptor P2X4 isoform a; purinergic receptor P2X4 subunit variant 1; purinergic receptor P2X4 subunit variant 2; purinergic receptor P2x4 subunit variant e; purinoceptor P2X4; putative partial extracellular loop region | |
Goat | |
Antigen affinity chromatography | |
RUO | |
5025 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q99571 | |
P2RX4 | |
Peptide with sequence C-ETDSVVSSVTTKAK, from the internal region of mouse P2rx4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |