missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA525581
Code : LD
Additional Details : Weight : 0.01000kg
Description
P2X7 Polyclonal antibody specifically detects P2X7 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
P2X7 | |
Polyclonal | |
Unconjugated | |
P2RX7 | |
AI467586; ATP receptor; MGC20089; P2rx7; P2X purinoceptor 7; p2X receptor 7; P2X(7); P2X7; P2X7 purinoceptor; P2X7 receptor; P2X7R; P2Z; P2Z receptor; Purinergic receptor; purinergic receptor P2X 7; purinergic receptor P2X, ligand gated ion channel, 7; purinergic receptor P2X, ligand-gated ion channel, 7; purinergic receptor P2X7; purinergic receptor P2X7 variant A | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
18439, 5027 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q99572, Q9Z1M0 | |
P2RX7 | |
KLH conjugated synthetic peptide between 563-592 amino acids from the C-terminal region of human P2RX7 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |