missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-p27Kip1, Polyclonal, Epredia™ Lab Vision™
Brand: Epredia RB-9019-P
Code : LD
Additional Details : Weight : 0.00100kg
Specifications
p27Kip1 | |
200μg/mL | |
Rabbit | |
1 mL | |
Cancer and Tumor Biology | |
Human, Mouse, Rat | |
IgG |
Polyclonal | |
Unconjugated | |
Synthetic peptide derived from C-terminal of human p27Kip1 | |
RUO | |
Primary | |
Concentrated |