missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-p27Kip1, Polyclonal, Epredia™ Lab Vision™
Brand: Epredia RB-9019-R7
Code : LD
Additional Details : Weight : 0.02000kg
Specifications
p27Kip1 | |
Polyclonal | |
Unconjugated | |
Synthetic peptide derived from C-terminal of human p27Kip1 | |
RUO | |
Primary | |
Prediluted |
Polyclonal | |
n/a | |
Rabbit | |
7 mL | |
Cancer and Tumor Biology | |
Human, Mouse, Rat | |
IgG |