missing translation for 'onlineSavingsMsg'
Learn More
Learn More
K-Ras Monoclonal Antibody (F132-62), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA191046
Code : LE
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Sw480 or Y1 cells or normal skin tissue..
K-Ras Monoclonal antibody specifically detects K-Ras in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation.Specifications
K-Ras | |
Monoclonal | |
0.1 mg/mL | |
0.05M sodium phosphate with 0.2% gelatin and 0.1% sodium azide | |
P01116, P08644, P32883 | |
KRAS | |
Recombinant full length human protein. | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation | |
F132-62 | |
Unconjugated | |
KRAS | |
AI929937; cellular c-Ki-ras2; cellular c-Ki-ras2 proto-oncogene; CFC2; c-Ki-ras; c-Kirsten-ras protein; c-Kirsten-ras proto-oncogene; c-K-ras; c-K-ras2 protein; fa04e08; fc14b12; fc23g10; fj89d12; GTPase KRas; GTPase KRas, N-terminally processed; hypothetical protein LOC445289; K RAS; ki-Ras; Kirsten rat sarcoma oncogene 2, expressed; Kirsten rat sarcoma viral (Kras-2) oncogene homolog; Kirsten rat sarcoma viral oncogene; Kirsten rat sarcoma viral oncogene homolog; Kirsten rat sarcoma viral oncogene homologue 2 (active); KRAS; K-ras; K-Ras 2; K-ras p21 protein; KRAS proto-oncogene, GTPase; KRAS1; Kras2; Kras-2; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; NS; NS3; oncogene KRAS2; p21; p21 protein; p21B; p21ras; PR310 c-K-ras oncogene; RALD; ras; ras p21; RASK; RASK2; ras-like protein; transforming protein p21; Unknown; v-Ki-ras2 Kirsten rat sarcoma 2 viral oncogene homolog; v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog; wu:fa04e08; wu:fc14b12; wu:fc23g10; wu:fj89d12; xRAS2; zgc:85725 | |
Mouse | |
Protein A | |
RUO | |
16653, 24525, 3845 | |
4° C, do not freeze | |
Liquid |