missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p21 Monoclonal Antibody (WA-1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA133926
Code : LE
Additional Details : Weight : 0.01000kg
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
p21 Monoclonal antibody specifically detects p21 in Human, Non-human Primate samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
p21 | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide | |
P38936 | |
CDKN1A | |
Full length native protein (purified) (Human). | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
WA-1 | |
Unconjugated | |
CDKN1A | |
Activating Fragment 1; CAP20; cdk inhibitor CIP1; cdk inhibitor CIP1 (p21); CDKI; CDK-interacting protein 1; CDK-interaction protein 1; Cdkn1; CDKN1A; CDNK1A; CIP1; Cip1); cyclin dependent kinase inhibitor 1A; cyclin-dependent kinase inhibitor 1; cyclin-dependent kinase inhibitor 1A; Cyclin-dependent kinase inhibitor 1A (p21; cyclin-dependent kinase inhibitor 1A (P21); cyclin-dependent kinase inhibitor 1A (p21, Cip1); cyclin-dependent kinase inhibitor 1A variant 1; cyclin-dependent kinase inhibitor 1A variant 2; DNA synthesis inhibitor; MDA6; MDA-6; melanoma differentiation associated protein 6; melanoma differentiation-associated protein; Melanoma differentiation-associated protein 6; OTTHUMP00000016298; p21; p21<WAF1/CIP1>; p21Cip1; p21Cip1/Waf1; p21WAF; PIC1; putative cyclin-dependent kinase inhibitor 1A (p21/WAF1/CIP1); SDI1; SLC12A9; UV96; WAF1; Wild type p53 activated fragment 1 (WAF1); wild-type p53-activated fragment 1 | |
Mouse | |
Protein A | |
RUO | |
1026 | |
4° C, do not freeze | |
Liquid |