Learn More
p16INK4a Monoclonal Antibody (1E12E10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517142
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17142 targets P16 in indirect ELISA, IHC and WB applications and shows reactivity with Human, Mouse, and Rat samples. The MA5-17142 immunogen is purified recombinant fragment of P16 expressed in E. Coli.
p16INK4a Monoclonal antibody specifically detects p16INK4a in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot.Specifications
p16INK4a | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P42771, Q64364, Q8QZZ9 | |
CDKN2A | |
Purified recombinant fragment of P16 expressed in E. Coli. | |
RUO | |
1029, 12578, 25163 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1E12E10 | |
Unconjugated | |
CDKN2A | |
Alternative reading frame; Arf; ARF-INK4a; CDK4 inhibitor p16 INK4; CDK4 inhibitor p16-INK4; CDK4I; CDKN2; Cdkn2a; cdkn2a {ECO:0000312; cell cycle inhibitor; cell cycle negative regulator beta; cell cycle regulator; CMM2; Cyclin dependent kinase 4 inhibitor A; cyclin dependent kinase inhibitor 2A; Cyclin dependent kinase inhibitor 2A (p16, inhibits CDK4); cyclin-dependent kinase 4 inhibitor A; Cyclin-dependent kinase inhibitor 2A; cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2A (p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2a p16Ink4a; cyclin-dependent kinase inhibitor 2a p19Arf; cyclin-dependent kinase inhibitor 2A, isoform 1; cyclin-dependent kinase inhibitor 2A, isoform 2; cyclin-dependent kinase inhibitor 2A, isoform 3; cyclin-dependent kinase inhibitor 2A, isoforms 1/2; cyclin-dependent kinase inhibitor protein; EMBL:AAL76336.1, ECO:0000312; EMBL:AAL76338.1, ECO:0000312; HGNC:1787; inhibits CDK4; INK4; INK4A; Ink4 | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |