Learn More
Otx2 Mouse anti-Human, Clone: 246826, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1979
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13469075
  • £288.60 / 100µg
Estimated Shipment
14-08-2020
Description
Description
Otx2 Monoclonal antibody MAB1979 Specifically detects Otx2 in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications
Specifications
Otx2
246826
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
OTX2
E. coli-derived recombinant human Otx2 (isoform b) Met1-Leu289 Accession # P32243
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5015
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunocytochemistry
LYOPH
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
P32243
homeobox protein OTX2, MCOPS5, MGC45000, orthodenticle homeobox 2, Orthodenticle homolog 2, orthodenticle homolog 2 (Drosophila), OTX2
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Otx2 in Western blots.