missing translation for 'onlineSavingsMsg'
Learn More

Osteoprotegerin/TNFRSF11B Rat anti-Mouse, Clone: 110511, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB4591

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460626

  • £337.93 / 500µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Osteoprotegerin/TNFRSF11B Monoclonal antibody MAB4591 Specifically detects Osteoprotegerin/TNFRSF11B in Mouse samples. It is validated for ELISA Capture, Western Blotting
Specifications

Specifications

Osteoprotegerin/TNFRSF11B
110511
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Q6PI12
OCIF, OCIFMGC29565, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TNFRSF11B, TR1, tumor necrosis factor receptor superfamily, member 11b
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Osteoprotegerin/TNFRSF11B in ELISAs and Western blots. In Western blots, this antibody shows 100% cross-reactivity with recombinant human (rh) Osteoprotegerin and no cross-reactivity with rh4-1BB, recombinant mouse (rm) CD27, rmCD40, rhDR3, rhDR6, rmFas, rmGITR, rhHVEM, rhLT beta R, rmRANK, rmTNF RI, or rmTNF RII.
Mouse
Western Blot, ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TNFRSF11B
Mouse myeloma cell line NS0-derived recombinant mouse Osteoprotegerin/TNFRSF11B Gln21-Leu401 (Gln138Arg) Accession # Q6PI12
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4982
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos

For Research Use Only