missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSBP Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518364
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology.
OSBP Polyclonal antibody specifically detects OSBP in Human samples. It is validated for Western Blot.Specifications
OSBP | |
Polyclonal | |
Unconjugated | |
Osbp | |
1110018F06Rik; AW559088; HGNC:8503; Kiaa4220; mKIAA4220; OSBP; OSBP1; oxysterol binding protein; oxysterol-binding protein 1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
5007 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P22059 | |
Osbp | |
Synthetic peptide sequence (KHQLEETKKEKRTRIP) corresponding to the internal amino acids of OSBP | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |