Learn More
ORC4 Monoclonal Antibody (2A8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA519508
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: MSSRKSKSNS LIHTECLSQV QRILRERFCR QSPHSNLFGV QVQYKHLSEL LKRTALHGES NSVLIIGPRG SGKTMLISHA LKELMEIEEV SENVLQVHLN GLLQINDKIA LKEITRQLNL ENVVGDKVFG SFAENLSFLL EALKKGDRTS SCPVIFILDE FDLFAHHKNQ TLLYNLFDIS QSAQTPIAVI GLTCRLDILE LLEKRVKSRF SHRQIHLMNS FGFPQYVKIF KEQLSLPAEF PDKVFAEK
ORC4 Monoclonal antibody specifically detects ORC4 in Human samples. It is validated for ELISA, Immunocytochemistry.Specifications
ORC4 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
O43929 | |
ORC4 | |
orc4 (AAH05388, 1 a.a. ∽ 248 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Immunocytochemistry | |
2A8 | |
Unconjugated | |
ORC4 | |
HSORC4; mMmORC4; Orc4; ORC4L; Orc4P; Origin recognition complex subunit 4; origin recognition complex, subunit 4; origin recognition complex, subunit 4 homolog; origin recognition complex, subunit 4-like | |
Mouse | |
Affinity chromatography | |
RUO | |
5000 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |