missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523390
Code : LD
Additional Details : Weight : 0.01000kg
Description
ORC1 Polyclonal antibody specifically detects ORC1 in Human, Mouse samples. It is validated for Western Blot.Specifications
ORC1 | |
Polyclonal | |
Unconjugated | |
Orc1 | |
AA545195; HS; HSORC1; MmORC1; orc1; orc1.L; Orc1l; origin recognition complex protein 1; origin recognition complex subunit 1; origin recognition complex subunit 1 L homeolog; origin recognition complex, subunit 1; origin recognition complex, subunit 1 homolog; origin recognition complex, subunit 1-like; PARC1; Replication control protein 1; XELAEV_18023178mg; xorc1 | |
Rabbit | |
Protein A | |
RUO | |
18392, 4998 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
Q13415, Q9Z1N2 | |
Orc1 | |
A portion of amino acids 170 to 220 of human ORC1L | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |