missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR2A4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534048
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human (100%) Horse (93%) Marmoset, Panda, Dog, Opossum (86%).
OR2A4 Polyclonal antibody specifically detects OR2A4 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
OR2A4 | |
Polyclonal | |
Unconjugated | |
OR2A4 | |
Olfactory receptor 2A10; olfactory receptor 2A4; olfactory receptor family 2 subfamily A member 4; olfactory receptor OR6-37; olfactory receptor, family 2, subfamily A, member 10; olfactory receptor, family 2, subfamily A, member 4; OR2A10; OR2A4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79541 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O95047 | |
OR2A4 | |
Synthetic 14 amino acid peptide from 2nd extracellular domain of human OR2A4. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |