Learn More
Oncostatin M/OSM Rat anti-Mouse, Clone: 157210, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB495
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13450776
  • £405.72 / 500µg
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
Oncostatin M/OSM Monoclonal antibody MAB495 Specifically detects Oncostatin M/OSM in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Oncostatin M/OSM
157210
Western Blot 1 ug/mL
P53347.1
MGC20461, oncostatin M, oncostatin-M
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse OSM in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant hhuman (rh) OSM, rhCLC, recombinant mouse (rm) CT-1, rmIL-6, rmIL-11, rmLIF, or recombinant rat CNTF.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
OSM
E. coli-derived recombinant mouse OSM Ala24-Arg206 Accession # P53347.1
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5008
Reconstitute at 0.5 mg/mL in sterile PBS.