missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA536258
Code : NEW
Additional Details : Weight : 0.00910kg
Description
This antibody detects endogenous protein at a molecular weight of 45 kDa. Purity is >95% by SDS-PAGE.
OCT6 Polyclonal antibody specifically detects OCT6 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
OCT6 | |
Polyclonal | |
Unconjugated | |
POU3F1 | |
6-Oct; LOW QUALITY PROTEIN: POU domain, class 3, transcription factor 1; Oct6; Oct-6; octamer binding factor oct6; octamer-binding protein 6; Octamer-binding transcription factor 6; octoamer transcription factor-6; octomer-6; Otf6; OTF-6; POU class 3 homeobox 1; POU domain transcription factor SCIP; POU domain, class 3, transcription factor 1; POU3F1; Scip; suppressed cAMP-inducible POU; Test1; Testes-1; Tst1; Tst-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18991, 192110, 5453 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P20267, P21952, Q03052 | |
POU3F1 | |
Synthetic peptide corresponding to amino acids 300-350 of human Oct-6 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |