Learn More
OCAM/NCAM2 Rat anti-Mouse, Clone: 125101, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB778
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13461856
  • £337.93 / 500µg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
OCAM/NCAM2 Monoclonal antibody MAB778 Specifically detects OCAM/NCAM2 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
OCAM/NCAM2
125101
Western Blot 1 ug/mL
O35136
NCAM2, N-CAM-2, NCAM-2, NCAM21MGC51008, neural cell adhesion molecule 2, neural cell adhesion molecule 2 precursor10, Rb-8, RNCAM
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse OCAM/NCAM2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ALCAM, rhBCAM, rhEpCAM, rhMCAM, recombinant mouse (rm) NCAM-L1, or rmMAdCAM-1 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
NCAM2
Mouse myeloma cell line NS0-derived recombinant mouse OCAM/NCAM2 Leu20-Gly700 (predicted) Accession # O35136
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4685
Reconstitute at 0.5 mg/mL in sterile PBS.