missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OAZ1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA514385
Code : LE
Additional Details : Weight : 0.01000kg
Description
OAZ1 Polyclonal antibody specifically detects OAZ1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
OAZ1 | |
Polyclonal | |
Unconjugated | |
OAZ1 | |
Antizyme; antizyme 1; AZ1; AZ-1; AZI; OAZ; OAZ1; ODCAC; ODC-Az; Ornithine decarboxylase antizyme 1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
18245, 4946 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P54368, P54369 | |
OAZ1 | |
KLH conjugated synthetic peptide between 2-31 amino acids from the N-terminal region of human OAZ1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |