Learn More
NT-4 Mouse anti-Human, Clone: 36507, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2681
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439625
  • £390.72 / 500µg
Estimated Shipment
14-09-2020
Description
Description
NT-4 Monoclonal antibody MAB2681 Specifically detects NT-4 in Human samples. It is validated for Western Blotting
Specifications
Specifications
NT-4
36507
Western Blot 1 ug/mL
P34130
GLC10, GLC1O, neurotrophic factor 4, neurotrophic factor 5, neurotrophin 4, neurotrophin 5 (neurotrophin 4/5), neurotrophin-4, neurotrophin-5, Neutrophic factor 4, NT4, NT-4, NT-4/5, NT-5, NTF4, NTF5, NTF5NT5
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human NT-4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) BDNF or rhNT-3 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
NTF4
E. coli-derived recombinant human NT-4 Gly81-Ala210 (predicted) Accession # P34130
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4909
Reconstitute at 0.5 mg/mL in sterile PBS.