missing translation for 'onlineSavingsMsg'
Learn More

NTF3, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA118391

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11572032

  • £297.00 / 500µg


Estimated Shipment
22-06-2018
Add to basket

Description

Description

PA1-18391 detects NT3 from human, rat, and chicken samples.PA1-18391 has been successfully used in immunohistochemistry, ELISA, Western blot, and inhibition assay procedures.The PA1-18391 immunogen is recombinant human NT3.Reconstitute with 500 ul of distilled water.
Specifications

Specifications

Neurotrophin-3
Polyclonal
Unconjugated
NTF3
Purified
NTF3
Recombinant human NT3.
Protein G
store at -20°C, Avoid Freeze/Thaw Cycles
Primary
4908
ELISAþImmunohistochemistryþInhibition AssaysþWestern Blot
1mg/mL
Antibody with no preservative
P20783, P18280
Neurotrophin-3, Neurotrophic factor, HDNF, Nerve growth factor 2, NGF-2, Ntf3, Ntf-3
Rabbit
IgG
500μg
Antibody
Polyclonal
HumanþNon-human PrimateþRat
Videos