Learn More
NPR-1 Rat anti-Mouse, Clone: 488620, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3974
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13460346
  • £249.49 / 100µg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
NPR-1 Monoclonal antibody MAB3974 Specifically detects NPR-1 in Mouse samples. It is validated for Immunohistochemistry
Specifications
Specifications
NPRA/NPR1
488620
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
NPR1
Mouse myeloma cell line NS0-derived recombinant mouse NPRA/NPR1 Ser29-Glu469 (predicted) Accession # P18293
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4881
Reconstitute at 0.5 mg/mL in sterile PBS.
Immunohistochemistry
LYOPH
Immunohistochemistry 8-25 ug/mL
P18293
ANPa, ANP-A, ANPRA, ANPR-A, ANPRAEC 4.6.1.2, atrial natriuretic peptide receptor 1, Atrial natriuretic peptide receptor type A, atrionatriuretic peptide receptor A, GC-A, Guanylate cyclase A, GUC2A, GUCY2A, natriuretic peptide A type receptor, natriuretic peptide receptor A, natriuretic peptide receptor A/guanylate cyclase A (atrionatriuretic peptidereceptor A), NPR1, NPRA, NPR-A, Pndr
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse NPRA/NPR1 in direct ELISAs.