missing translation for 'onlineSavingsMsg'
Learn More

NPC1-Like 1 Polyclonal Antibody, Invitrogen™

NPC1-Like 1 Antibody, PA1-16801, from Invitrogen™. Species Reactivity: Human, Mouse, Rat; Applications: Immunohistochemistry, Immunoprecipitation, Western Blot.

Brand:  Invitrogen PA1-16801

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11575383

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

Dietary consumption and intestinal cholesterol absorption contribute to plasma cholesterol levels, which factor into the risk of coronary heart disease. Niemann-Pick type C1 Like 1 protein (NPC1L1 or NPC3) is enriched in the small intestine and is found in the brush border membrane of enterocytes. It plays a critical role in absorption of intestinal cholesterol.
Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Isotype: IgG
Immunogen: A synthetic peptide made to an internal region of rat NPC1L1 (between residues 500-600).
Formulation: 100μL purified antibody (1mg/mL) in tris citrate and 0.01% sodium azide
Synonyms: NPC3, Niemann-Pick C3, NPC1L1, Niemann-Pick type C1 Like 1 Protein, BC 400-127
Uniprot Accession: Q9UHC9, Q6T3U4, Q6T3U3
Entrez GeneID: 29881, 237636, 432367
Gene Symbol: NPC1L1
Specifications

Specifications

NPC1-Like 1
Unconjugated
Q9UHC9, Q6T3U4, Q6T3U3
NPC3, Niemann-Pick C3, NPC1L1, Niemann-Pick type C1 Like 1 Protein, BC 400-127
Rabbit
IgG
Store at 4° C, do not freeze
Polyclonal
HumanþMurineþRat
ImmunohistochemistryþImmunoprecipitationþWestern Blot
100μL purified antibody (1mg/mL) in tris citrate and 0.01% sodium azide
Purified
NPC1L1
A synthetic peptide made to an internal region of rat NPC1L1 (between residues 500-600).
100μL
Primary
29881, 237636, 432367
Videos