missing translation for 'onlineSavingsMsg'
Learn More

NOV/CCN3 Rat anti-Mouse, Clone: 231226, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB19762

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459075

  • £337.93 / 500µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

NOV/CCN3 Monoclonal antibody MAB19762 Specifically detects NOV/CCN3 in Mouse samples. It is validated for ELISA Capture
Specifications

Specifications

NOV/CCN3
231226
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
NOV
Mouse myeloma cell line NS0-derived recombinant mouse NOV/CCN3, Ser26-Ile354, Accession # Q64299
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4856
Reconstitute at 0.5 mg/mL in sterile PBS.
ELISA
LYOPH
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Q64299
CCN family member 3, CCN3, CCN3IGF-binding protein 9, IBP-9, IGFBP-9, IGFBP9novH, Insulin-like growth factor-binding protein 9, nephroblastoma overexpressed gene, Nephroblastoma-overexpressed gene protein homolog, NOVH, protein NOV homolog
Rat
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects mouse NOV/CCN3 in ELISAs.
Mouse
Videos

For Research Use Only