Learn More
Norovirus capsid G2 Monoclonal Antibody (L34D), Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA518241
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This antibody does not cross-react with capsid G1. Purity is >90%. Recommended pairs for ELISA: MA5-18240 (capture) and MA5-18241 (conjugated, detection) or MA5-18241 (capture) and MA5-18242 (conjugated, detection). This antibody can also be used as a capture antibody in lateral flow applications.
Norovirus capsid G2 Monoclonal antibody specifically detects Norovirus capsid G2 in Virus samples. It is validated for ELISA, Lateral Flow, Western Blot.Specifications
Norovirus capsid G2 | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2b |
ELISA, Lateral Flow, Western Blot | |
L34D | |
Unconjugated | |
Norwalk virus | |
Norovirus recombinant protein | |
100 μg | |
Primary | |
4° C | |
Liquid |