missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOL8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535937
Code : NEW
Additional Details : Weight : 0.00910kg
Description
Recommended positive controls: HeLa, HeLa nuclear. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NOL8 Polyclonal antibody specifically detects NOL8 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
NOL8 | |
Polyclonal | |
Unconjugated | |
NOL8 | |
4921532D18Rik; 5730412B09Rik; bA62C3.3; bA62C3.4; C9orf34; NOL8; NOP132; Nucleolar protein 8; nucleolar protein Nop132 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55035 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q76FK4 | |
NOL8 | |
Recombinant fragment corresponding to a region within amino acids 150 and 280 of NOL8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |