Learn More
NOD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521981
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Raji. Predicted reactivity: Mouse (81%), Rat (82%), Pig (87%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NOD1 Polyclonal antibody specifically detects NOD1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
NOD1 | |
Polyclonal | |
Unconjugated | |
NOD1 | |
C230079P11; Card4; caspase recruitment domain 4; caspase recruitment domain family, member 4; caspase recruitment domain-containing protein 4; CLR7.1; F830007N14Rik; NLR family, CARD domain containing 1; Nlrc1; Nod1; nucleotide binding oligomerization domain containing 1; nucleotide-binding oligomerization domain containing 1; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 1; nucleotide-binding oligomerization domain-containing protein 1; RGD1562269 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10392 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y239 | |
NOD1 | |
Recombinant fragment corresponding to a region within amino acids 698 and 953 of NOD1 (Uniprot ID#Q9Y239) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |