missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDAR2D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA536539
Code : NEW
Additional Details : Weight : 0.00910kg
Description
This antibody detects endogenous protein at a molecular weight of 170 kDa. Purity is >95% by SDS-PAGE.
NMDAR2D Polyclonal antibody specifically detects NMDAR2D in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
NMDAR2D | |
Polyclonal | |
Unconjugated | |
GRIN2D | |
EB11; estrogen receptor binding CpG island; GluN2D; glur epsilon 4; GluRepsilon4; glutamate [NMDA] receptor subunit epsilon-4; glutamate ionotropic receptor NMDA type subunit 2D; glutamate receptor; glutamate receptor ionotropic, NMDA 2D; glutamate receptor, ionotropic, NMDA2D; glutamate receptor, ionotropic, NMDA2D (epsilon 4); glutamate receptor, ionotropic, N-methyl D-aspartate 2D; GRIN2D; ionotropic NMDA glutamate receptor epsilon 4 subunit type 2; NMDAR2D; N-methyl D-aspartate receptor subtype 2D; N-methyl-d-aspartate receptor subunit 2D; NR2D | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14814, 24412, 2906 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide; pH 7.2 | |
O15399, Q03391, Q62645 | |
GRIN2D | |
Synthetic peptide corresponding to amino acids 661-710 of human NMDA epsilon4 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |