Learn More
NMDAR2B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA34665
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is specific for the ∽180 kDa NR2B subunit of the NMDA Receptor. There is no reactivity towards the NR2A and NR2C subunits. Immunolabeling is blocked by preadsorption of antibody with the fusion protein used to generate the antibody.
NMDAR2B Polyclonal antibody specifically detects NMDAR2B in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
NMDAR2B | |
Polyclonal | |
Unconjugated | |
GRIN2B | |
AW490526; EIEE27; GluN2B; GluRepsilon2; glutamate [NMDA] receptor subunit epsilon-2; glutamate ionotropic receptor NMDA type subunit 2B; glutamate receptor; glutamate receptor ionotropic, NMDA 2B; glutamate receptor subunit epsilon-2; glutamate receptor, ionotropic, NMDA2B; glutamate receptor, ionotropic, NMDA2B (epsilon 2); glutamate receptor, ionotropic, N-methyl D-aspartate 2B; GRIN 2B; GRIN2B; HGNC:4586; hNR 3; hNR3; LOW QUALITY PROTEIN: glutamate receptor ionotropic, NMDA 2B; MGC142178; MGC142180; MRD6; Nmdar2b; NMDE2; NME2; N-methyl D-aspartate receptor 2B; N-methyl D-aspartate receptor subtype 2B; N-methyl-D-aspartate receptor subunit 2B; N-methyl-D-aspartate receptor subunit 3; NR2B; NR3 | |
Rabbit | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
Conc. Not Determined | |
whole serum with no preservative | |
Q00960, Q01097, Q13224 | |
GRIN2B | |
Fusion protein from the C-terminal region of the NR2B subunit of rat GRIN2B | |
RUO | |
14812, 24410, 2904 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |