Learn More
NKp44/NCR2 Mouse anti-Human, Clone: 253422, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2249
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13459295
  • £249.49 / 100µg
Estimated Shipment
06-03-2019
Add to basket
Description
Description
NKp44/NCR2 Monoclonal antibody MAB2249 Specifically detects NKp44/NCR2 in Human samples. It is validated for Western Blotting
Specifications
Specifications
NKp44/NCR2
253422
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
NCR2
Mouse myeloma cell line NS0-derived recombinant human NKp44/NCR2 Gln22-Pro190 Accession # O95944
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
9436
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 2 ug/mL
O95944
CD336, CD336 antigen, LY95, LY95lymphocyte antigen 95 homolog (activating NK-receptor; NK-p44), lymphocyte antigen 95 (activating NK-receptor; NK-p44), Lymphocyte antigen 95 homolog, natural cytotoxicity triggering receptor 2, Natural killer cell p44-related protein, NCR2, NK cell activating receptor (NKp44), NK cell-activating receptor, NKP44, NK-p44dJ149M18.1
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human NKp44/NCR2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse MAR-1, recombinant human (rh) NKp30, rhNKp46, or rhNKp80 is observed.