missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKG2C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528837
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: K562. Predicted reactivity: Rhesus Monkey (100%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NKG2C Polyclonal antibody specifically detects NKG2C in Human samples. It is validated for Western Blot.Specifications
NKG2C | |
Polyclonal | |
Unconjugated | |
KLRC2 | |
CD159 antigen-like family member C; CD159A; CD159c; cd159e; killer cell lectin like receptor C2; killer cell lectin-like receptor subfamily C, member 2; KLRC1; KLRC2; NK cell receptor C; NKG2; NKG2A; NKG2C; NKG2-C; NKG2-C type II integral membrane protein; NKG2-C-activating NK receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3822 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.56 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P26717 | |
KLRC2 | |
Synthetic peptide corresponding to a region within amino acids 1 and 45 of NKG2-C | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |