missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517574
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
NIK Polyclonal antibody specifically detects NIK in Bovine, Equine, Human, Mouse, Non-human Primate, Rat samples. It is validated for Western Blot.Specifications
NIK | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
Q99558, Q9WUL6 | |
MAP3K14 | |
Synthetic peptide corresponding to residues adjacent to glycine 659 of human NIK | |
100 μL | |
Primary | |
Bovine, Equine, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MAP3K14 | |
aly; alymphoplasia; FTDCR1B; HS; HsNIK; Map3k14; Mitogen-activated protein kinase kinase kinase 14; NF kappa B-inducing kinase; NF-kappa-beta-inducing kinase; Nfkb inducing kinase; NIK; protein kinase; serine/threonine-protein kinase NIK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100050140, 360640, 508367, 53859, 9020 | |
-20°C | |
Liquid |