Learn More
NgR2/NgRH1 Mouse anti-Human, Clone: 333413, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2776
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439685
  • £249.49 / 100µg
Estimated Shipment
09-07-2019
Description
Description
NgR2/NgRH1 Monoclonal antibody MAB2776 Specifically detects NgR2/NgRH1 in Human samples. It is validated for Western Blotting
Specifications
Specifications
NgR2/NgRH1/RTN4RL2
333413
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
RTN4RL2
Mouse myeloma cell line NS0-derived recombinant human NgR2/NgRH1 Ala28-Leu401 Accession # Q86UN3
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
349667
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q86UN3
NgR2, NGRH1, reticulon 4 receptor-like 2, RTN4RL2
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human NgR2/NgRH1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) NgR3 or rhNgR is observed.