Learn More
NGF Monoclonal Antibody (25623), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523717
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) beta -NGF and recombinant rat (rr) beta &#x2011;NGF is observed and less than 5% cross-reactivity with recombinant human (rh) BDNF, rhNT-3, and rhNT-4 is observed. This antibody neutralizes the bioactivity of rh beta -NGF, rm beta -NGF, and rr beta -NGF. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
NGF Monoclonal antibody specifically detects NGF in Human samples. It is validated for ELISA, Neutralization.Specifications
NGF | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P01138 | |
NGF | |
Mouse myeloma cell line NS0-derived recombinant human beta-NGF Ser122-Ala241 | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Neutralization | |
25623 | |
Unconjugated | |
NGF | |
B nerve growth factor; beta ngf; beta-nerve growth factor; BetaNGF; beta-NGF; bNGF; b-NGF; HSAN5; MGC161426; MGC161428; nerve growth factor; nerve growth factor (beta polypeptide); nerve growth factor, beta; nerve growth factor, beta polypeptide; nerve growth factor, beta subunit; NGF; NGF b; NGF β; Ngfb; NGFbeta; NGFβ; β nerve growth factor; β NGF; βNGF | |
Mouse | |
Protein A/G | |
RUO | |
4803 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |