missing translation for 'onlineSavingsMsg'
Learn More

NGAL, clone: 17, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen ABS0471702

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15386291

  • £555.01 / 200µL


Estimated Shipment
04-07-2018
Add to basket

Description

Description

Reactivity: ABS 047-17-02 binds free canine NGAL. ABS 047-17-02 can be used as a capture antibody in a sandwich ELISA using biotinylated detection antibody ABS 047-14-02, giving a detection limit of 5 pg/mL in an unoptimized buffer assay.NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/ml
Specifications

Specifications

NGAL/LCN2
Monoclonal
1mg/mL
PBS with 15mM sodium azide; pH 7.4
Purified
LCN2
Recombinant dog NGAL adsorbed onto aluminum hyroxide gel.
Protein A
Store at 4° C, store in dark
Primary
491320
ELISAþWestern Blot
17
Unconjugated
LCN2
Neutrophil gelatinase-associated lipocalin, HNL, p25, 25kDa alpha-2-microglobulin-related subunit of MMP-9, Lipocalin-2, Oncogene 24p3
Murine
IgG2a, κ
200μL
Antibody
Monoclonal
Canine
Videos